EMAIL THIS PAGE TO A FRIEND
E3273 Sigma

Endomorphin 1

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Opioid Peptides, Other Opioid Peptides,
InChI Key   ZEXLJFNSKAHNFH-SYKYGTKKSA-N
storage temp.   −20°C
Gene Information   human ... OPRM1(4988)
mouse ... OPRM1(18390)
rat ... OPRM1(25601)

Description

Amino Acid Sequence

Tyr-Pro-Trp-Phe-NH2

Biochem/physiol Actions

Potent, selective endogenous μ opioid receptor agonist.

Legal Information

Sold under license to US patent number 6,303,578

Other Notes

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. E3273.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?