• USA Home
  • E3506 - [des-Tyr1]-Leucine enkephalinamide acetate salt

EMAIL THIS PAGE TO A FRIEND
E3506 Sigma

[des-Tyr1]-Leucine enkephalinamide acetate salt

≥95% (HPLC)

Synonym: Gly-Gly-Phe-Leu-NH2

  • CAS Number 100929-57-5

  • Empirical Formula (Hill Notation) C19H29N5O4

  • Molecular Weight 391.46

  •  MDL number MFCD00133259

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Enkephalins, Opioid Peptides, Peptides and Proteins,
InChI Key   BIPXJRSFBIBQNG-YYLIZZNMSA-N
assay   ≥95% (HPLC)
storage temp.   −20°C

Price and Availability


Detect Your Unseen Protein

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?