• USA Home
  • E4397 - 5′-(N-Ethylcarboxamido)adenosine-[adenine-2,8-3H]

EMAIL THIS PAGE TO A FRIEND
E4397 Sigma

5′-(N-Ethylcarboxamido)adenosine-[adenine-2,8-3H]

≥95% (Radiochemical Purity), aqueous ethanol solution

Synonym: NECA

Purchase

Properties

assay   ≥95% (Radiochemical Purity)
form   aqueous ethanol solution
extent of labeling   20-40 Ci/mmol
storage temp.   −20°C

Description

Packaging

Serum bottle

Physical form

Ethanol: water (1:1) solution

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2910 7
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?