• USA Home
  • E4501 - N-Ethyl-5-phenylisoxazolium-3′-sulfonate

EMAIL THIS PAGE TO A FRIEND
E4501 Sigma

N-Ethyl-5-phenylisoxazolium-3′-sulfonate

~95%

Synonym: Woodward’s reagent K

Purchase

Properties

assay   ~95%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?