• USA Home
  • E4767 - Monoclonal Anti-S1P2, C-Terminal antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
E4767 Sigma

Monoclonal Anti-S1P2, C-Terminal antibody produced in mouse

clone AS65, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-EDG-5, Anti-Endothelial Cell Differentiation Gene 5, C-Terminal

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Lipids in Cell Signaling, Primary Antibodies,
species reactivity   human, rat
application(s)   western blot: suitable
clone   AS65, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG
mol wt   antigen mol wt ~45 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... S1PR2(9294)
rat ... S1pr2(29415)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

human EDG-5/S1P2 unique C-terminal peptide.

Antigen Background

EDG-5/S1P2 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. The ligand for EDG-5/S1P2 is sphingosine 1-phosphate, and is expressed in cardiovascular, central nervous system, gonadal, placental and leukocyte-containing tissue.

Preparation Note

Purified from in vitro cell culture supernatants using protein A.

Application

Monoclonal Anti-S1P2, C-Terminal antibody produced in mouse is suitable for western blotting at a working dilution of 1:1000 (1μg/mL) using human T lymphoblasts, ovarian tumor cells, epithelial cells, estrogen receptor positive breast carcinoma cells, human spleen and rat spleen or lung.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

Sphingosine-1-phosphate receptor 2 (S1PR2 or S1P2) is a human gene encodes a G protein-coupled receptor. It binds the lipid signaling molecule sphingosine 1-phosphate (S1P) and regulates vascular permeability along with S1P1. Activation of S1P2R in endothelial cells increases vascular permeability. It is a bioactive lysophospholipid capable of inducing a wide spectrum of biological responses. S1P acts as an intercellular mediator by interacting with the endothelial differentiation gene (EDG)/S1P family of G protein-coupled receptors. S1P2 receptor is involved in S1P-induced platelet aggregation and Rho kinase activation. Type 2 diabetes patient responds to S1P.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?