• USA Home
  • E4903 - Sartorius eLINE® single channel electronic pipettor
EMAIL THIS PAGE TO A FRIEND
E4903 Sigma
Sartorius eLINE® single channel electronic pipettor
volume 10-300 μL, AC input 110 V, US 2-pin plug
Synonym: Biohit pipettes, electronic pipettes, pipettor
Purchase
Properties
Related Categories BioHit Pipettors, Labware, Liquid Handling Equipment, Manufacturer Browser, Pipettes, Pipettors and Tips,
feature   electronic
  single-channel
mfr. no.   Sartorius, 730061
availability   not available in EU
volume   10-300 μL
Description
General description
Sartorius eLINE electronic pipettors of the Biohitfamily offer the latest technology in liquid handling. The eLINE electronic pipettor has been designed to deliver the highest levels of analytical accuracy with minimal the eLINE pipettors together with the ejection - take the effort out of pipetting in every laboratory.
The eLINE modes including:
• Forward Pipetting with Optional Cycle
• Reverse Pipetting
• Manual Pipetting
• Post Delivery Mixing
• Multi-Dispensing
• Multi-Aspirating
• Automated Multi-Dispensing
• Sequential-Dispensing
Legal Information
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?