EMAIL THIS PAGE TO A FRIEND
E5024 Sigma

EMB Agar

pH 7.2±0.2 (25 °C)

Synonym: Eosin Methylene Blue Agar

Purchase

Properties

pH   7.2±0.2 (25 °C)

Description

Application

Recommended for the isolation and differentiation of gram negative enteric bacteria from clinical and nonclinical specimens.

Other Notes

Components (g/L)
Peptic Digest of Animal Tissue 10.00
Dipotassium Phosphate 2.00
Lactose 5.00
Sucrose 5.00
Eosin-Y 0.40
Methylene Blue 0.065
Agar 13.50

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

70186 EMB Agar, for microbiology

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?