• USA Home
  • E5131 - [des-Tyr1, D-Ala2]-Methionine enkephalinamide acetate salt

EMAIL THIS PAGE TO A FRIEND
E5131 Sigma

[des-Tyr1, D-Ala2]-Methionine enkephalinamide acetate salt

≥95% (HPLC)

Synonym: D-Ala-Gly-Phe-Met-NH2

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   NXZJEQONDQLTQF-WEMUQIOZSA-N
assay   ≥95% (HPLC)
storage temp.   −20°C

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?