• USA Home
  • E5401 - p3XFLAG-CMV-18 Expression Vector

EMAIL THIS PAGE TO A FRIEND
E5401 Sigma

p3XFLAG-CMV-18 Expression Vector

Purchase

Properties

shipped in   dry ice
storage temp.   −20°C

Description

Application

A selectable vector for stable cytoplasmic expression of dual tagged (N-terminal Met-FLAG, C-terminal 6xHis) fusion proteins.

Other Notes

Stable expression is accomplished by neomycin selection (Geneticin, G-418).

Legal Information

p3xFLAG-CMV is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?