• USA Home
  • E6009 - Sigma-Aldrich® standard Ag/AgCl double junction pH combination electrode, glass body
EMAIL THIS PAGE TO A FRIEND
E6009 Sigma
Sigma-Aldrich® standard Ag/AgCl double junction pH combination electrode, glass body
BNC connector, electrode L 55 mm
Purchase
Properties
Related Categories Labware, pH Electrodes, pH Supplies More...
electrode L   55 mm
stem O.D.   8 mm
Description
General description
The combination of a pH indicating and Ag/AgCl reference half-cell in one electrode provides ease of handling and smaller sampling requirements. Combination double junction electrodes are Tris compatible with a maximum temperature of 80°C, and are recommended for proteins, metal ions, sulfides or other substances that will react with either Ag or AgCl. Combination Ag/AgCl electrodes are suggested for higher temperature applications (100°C). Both types supplied with a 30 in. lead.
Temperature Range:
• -5 to 100°C Continuous Use
• -5 to 130°C Intermittent Use
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
60137 Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?