• USA Home
  • E6139 - Big Endothelin 2 human

EMAIL THIS PAGE TO A FRIEND
E6139 Sigma

Big Endothelin 2 human

≥95% (HPLC)

  • CAS Number 132699-72-0

  • Empirical Formula (Hill Notation) C188H269N45O56S4

  • Molecular Weight 4183.68

  •  MDL number MFCD00131013

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Endothelins, Peptides and Proteins, Peptides for Cell Biology,
assay   ≥95% (HPLC)
storage temp.   −20°C
Gene Information   human ... EDN2(1907)

Description

Amino Acid Sequence

Cys-Ser-Cys-Ser-Ser-Trp-Leu-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-leu-Asp-Ile-Ile-Trp-Val-Asn-Thr-Pro-Glu-Gln-Thr-Ala-Pro-Tyr-Gly-Leu-Gly-Asn-Pro-Pro [Disulfide Bridges: 1-15; 3-11]

Price and Availability


Amplified Detection

Making Headway

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?