• USA Home
  • E6143 - Elastic TISSUE-TROL Control Slides

EMAIL THIS PAGE TO A FRIEND
E6143 Sigma

Elastic TISSUE-TROL Control Slides

from human skin containing elastic fibers

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, TISSUE-TROL Control Slides More...
Quality Level   GMP-IVD
biological source   from human skin containing elastic fibers
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Components

25 slides per box. 1 stained slide is provided with each box. The stained slide has been stained with Sigma Procedure No. HT25

Application

Suitable for use with Sigma Elastic Stain Procedure No. HT25A.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

TTR003 Elastic TISSUE-TROL Control Slides, from human skin containing elastic fibers

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?