EMAIL THIS PAGE TO A FRIEND
E6277 Sigma

β-Elastin bovine

lyophilized powder

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Signaling Enzymes, ECM Proteins, Elastin,
form   lyophilized powder
mol wt   mol wt 3-10 kDa
impurities   salt, free
solubility   H2O: soluble
storage temp.   2-8°C

Description

Preparation Note

Fractionated from α-elastin by method of Partrige, et al. Colloidal separation repeated two times to increase the purity of the β fractions. Uses E6527 as the starting material.

Application

â-Elastin is a structural protein which has been used in studies of chronic obstructive pulmonary disease (COPD). It may be used to investigate why there is a lack of repair for these proteins in patients with COPD.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?