• USA Home
  • E6389 - Big Endothelin 39 bovine

EMAIL THIS PAGE TO A FRIEND
E6389 Sigma

Big Endothelin 39 bovine

≥97% (HPLC)

  • CAS Number 124834-83-9

  • Empirical Formula (Hill Notation) C192H287N49O58S5

  • Molecular Weight 4369.95

  •  MDL number MFCD00131015

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥97% (HPLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-Val-Asn-Thr-Pro-Glu-His-Val-Val-Pro-Tyr-Gly-Leu-Gly-Ser-Pro-Ser-Arg-Ser [Disulfide Bridges: 1-15; 3-11]

Price and Availability


Amplified Detection

All labs need water

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?