EMAIL THIS PAGE TO A FRIEND
E6506 Sigma

Erucoyl coenzyme A

85-90%

Synonym: cis-13-Docosenoyl Coenzyme A

Purchase

Properties

assay   85-90%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?