• USA Home
  • E6650 - Anti-EphA1 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
E6650 Sigma

Anti-EphA1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Antibodies to Cytokines and Growth Factors,
species reactivity   human
application(s)   indirect ELISA: 0.5-1.0 μg/mL
  western blot: 0.1-0.2 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   human ... EPHA1(2041)
biological source   goat
conjugate   unconjugated

Description

Immunogen

recombinant human EphA1, extracellular domain, expressed in NSO cells.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline.

Specificity

By immunoblotting and ELISA, the antibody shows ~40% cross-reactivity with recombinant mouse EphA3, mouse EphA8, mouse EphA4, rat EphA5 and 10% cross-reactivity with recombinant mouse EphA2, mouse EphA6, and mouse EphA7.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?