EMAIL THIS PAGE TO A FRIEND
E6652 Sigma

α-Elastin from rat

powder

Purchase

Description

Preparation Note

Prepared from the elastin of mature rat lung by extraction utilizing coacervation as described by Partrige, et al.

Application

Elastin is a structural protein which has been used in studies of Chronic obstructive pulmonary disease (COPD). It may be used to investigate why there is a lack of repair for these proteins in patients with COPD.

The solubility of α-elastin has been applied to construction of elastin-mimetic biomaterials.

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?