• USA Home
  • E6883 - Elastase from porcine pancreas

EMAIL THIS PAGE TO A FRIEND
E6883 Sigma

Elastase from porcine pancreas

Type II-A, lyophilized powder, Protein ~70 %, ≥1 units/mg protein (biuret)

Synonym: Elastase from hog pancreas, Pancreatopeptidase E

Purchase

Description

Biochem/physiol Actions

Elastase hydrolyses elastin, the specific protein of elastic fibers, and digests hemoglobin, casein and fibrin.

Preparation Note

2× Crystallized

Unit Definition

One unit will hydrolyze 1.0 μmole of N-succinyl-L-Ala-Ala-Ala-p-nitroanilide per min, pH 8.0 at 25 °C.

Physical form

Contains sodium carbonate.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger, Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

inhibitor

Product #

Description

Add to Cart

M6159 α2-Macroglobulin from human plasma, BioUltra, ≥98% (SDS-PAGE)
D7910 3,4-Dichloroisocoumarin, serine protease inhibitor
D0879 Diisopropylfluorophosphate
A9024 α1-Antitrypsin from human plasma, salt-free, lyophilized powder
A6150 α1-Antitrypsin from human plasma, salt-free, lyophilized powder

substrate

Product #

Description

Add to Cart

M9771 N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin, elastase substrate
S4760 N-Succinyl-Ala-Ala-Ala-p-nitroanilide, elastase substrate
S8511 N-Succinyl-Ala-Ala-Pro-Leu p-nitroanilide, protease substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?