• USA Home
  • E7025 - Anti-EphB1 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
E7025 Sigma

Anti-EphB1 antibody produced in goat

affinity isolated antibody, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Antibodies to Cytokines and Growth Factors,
species reactivity   rat
application(s)   indirect ELISA: 0.5-1.0 μg/mL
  western blot: 0.1-0.2 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   rat ... Ephb1(24338)
biological source   goat
conjugate   unconjugated

Description

Immunogen

A recombinant rat EphB1 extracellular domain expressed in mouse NSO cells.

General description

Eph family of tyrosine kinase receptors mediates cell-cell attachment and aggregation by binding to specific ligands, ephrins. The Eph family consists of 14 structurally related receptors that are either membrane linked or are transmembrane with overlapping affinity for ligands. The transmembrane receptors EphA and EphB (ELK) subclasses signal via distinct mediators in the cytoplasm. EphB1 has several biological functions which includes neural synapsis formation and maturation, nervous system development and angiogenesis. It also interacts with the adaptor molecule Grb7 and regulates cell migration . Anti-EphB1 antibody can be used in western blotting. Goat anti-EphB1 antibody reacts specifically with recombinant rat EphB1. The antibody exhibits less than 5% (approx.) cross-reactivity with recombinant mouse EphB2, EphB3, EphB4, EphB6, EphA3, EphA4 and EphA7.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Specificity

By immunoblotting and ELISA, the antibody shows <5% cross-reactivity with recombinant mouse EphB2, mouse EphB3, mouse EphB4, mouse EphB6, mouse EphA3, mouse EphA4, and mouse EphA7.

Application

Anti-EphB1 antibody can be used in indirect ELISA (working concentration:- 0.5-1.0 μg/mL).

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?