• USA Home
  • E7647 - C-erbB-3 Fragment 1265-1278

EMAIL THIS PAGE TO A FRIEND
E7647 Sigma

C-erbB-3 Fragment 1265-1278

≥97% (HPLC)

Synonym: Ala-Ser-Glu-Gln-Gly-Tyr-Glu-Glu-Met-Arg-Ala-Phe-Gln-Gly

  • CAS Number 184901-83-5

  • Empirical Formula (Hill Notation) C67H99N19O25S

  • Molecular Weight 1602.68

  •  MDL number MFCD01310913

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
assay   ≥97% (HPLC)
storage temp.   −20°C

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?