EMAIL THIS PAGE TO A FRIEND
E8133 Sigma

Ethyl–Agarose

saline suspension, matrix 4% beaded agarose

Purchase

Properties

form   saline suspension
matrix   4% beaded agarose
matrix activation   epoxy
matrix attachment   amino
matrix spacer   12 atoms
capacity   4-6 mg/mL binding capacity (bovine serum albumin)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?