• USA Home
  • E8637 - (−)-Ephedrine hydrochloride

EMAIL THIS PAGE TO A FRIEND
E8637 Sigma

(−)-Ephedrine hydrochloride

drug standard, 1.0 mg/mL in methanol

Purchase

Properties

Related Categories Amphetamines / Stimulants Standards, Analytical Standards, Analytical/Chromatography, Pharmaceuticals, Illicit Drugs & Alcohol
grade   drug standard
assay   ≥98%
concentration   1.0 mg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Ephedrine Enantiomers on CHIRALPAK® AGP
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?