• USA Home
  • E8906 - Anti-EDEM3 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
E8906 Sigma

Anti-EDEM3 antibody produced in rabbit

~1.2 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym: Anti-ER degradation enhancer, mannosidase alpha-like 3

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Stress, Antibodies to Oxidative Stress Proteins,
species reactivity   rat, human, mouse
application(s)   immunoprecipitation: 2-5 μg using mouse 3T3 and human HepG2 cell lysates
  western blot: 2-5 μg/mL using whole extract of rat PC12 cells
clone   polyclonal
concentration   ~1.2 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt ~120 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... EDEM3(80267)
mouse ... Edem3(66967)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 470-485 of human EDEM3, conjugated to KLH. The corresponding sequence is identical in mouse and rat EDEM3.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Anti-EDEM3 antibody produced in rabbit is suitable for immunoprecipitation at a concentration of 2-5μg using mouse 3T3 and human HepG2 cell lysates and is also suitable for western blotting at a concentration of 2-5μg/mL using whole extract of rat PC12 cells.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

ER degradation-enhancing α-mannosidase-like 3 is an enzyme encoded by the EDEM3 gene in humans and consists of 931 amino acids. It consists of motifs of Class I α-mannosidases (glycosyl hydrolase family 47) in its N-terminal domain and a protease-associated motif in its C-terminal region. EDEM3 has α1,2-mannosidase activity. EDEM3 is also associated with folding-incompetent proteins carrying N-glycans that are extracted from futile folding cycles in the calnexin chaperone system.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

E8406 Anti-EDEM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
E8781 Anti-EDEM3 (C-terminal) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
E9906 Anti-EDEM2 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
E1782 Anti-EDEM2 (C-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?