• USA Home
  • E9135 - Exposure cassette, Kodak®
EMAIL THIS PAGE TO A FRIEND
E9135 Sigma
Exposure cassette, Kodak®
For 14 × 17 in and 35 × 43 cm film
Purchase
Properties
material   cardboard
  metal clip closure
Description
Caution
Recommended only for direct exposures at ambient temperatures, unless the holders are sealed in plastic wrap to prevent absorption of moisture.
General description
An inner envelope folds around the film to form a light-tight closure for autoradiography. The hinged outer cardboard covers are locked over the inner envelope with an attached metal clip.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?