-
Product Directory
Custom Product
-
Services Offered
Custom Capabilities
-
CAS Number 120796-97-6
Empirical Formula (Hill Notation) C189H282N48O56S5
Molecular Weight 4282.87
MDL number MFCD00081408
| Related Categories | Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, Cell Signaling and Neuroscience, |
| assay | ≥95% (HPLC) |
| storage temp. | −20°C |
| Gene Information |
Research your gene in Your Favorite Gene powered by Ingenuity human ... EDN1(1906) |
Cys-Ser-Cys-Ser-Ser-Leu-M
Precursor of endothelin 1; vasoconstrictor activity appears to be due to its conversion to endothelin 1 by endothelin converting enzyme in the tissue.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?