• USA Home
  • E9652 - EphB4/Fc Chimera from mouse

EMAIL THIS PAGE TO A FRIEND
E9652 Sigma

EphB4/Fc Chimera from mouse

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: Htk, Mdk2, Myk1, Tyro11

Purchase

Properties

Related Categories Angiogenesis, Application Index, Biochemicals and Reagents, Cancer Research, Cell Biology,
recombinant   expressed in NSO cells
assay   >95% (SDS-PAGE)
form   lyophilized powder
mol wt   monomer calculated mol wt 85.2 kDa
  mol wt ~110 kDa by SDS-PAGE (reducing)
impurities   endotoxin, tested
storage temp.   −20°C
Gene Information   mouse ... Ephb4(13846)

Description

Analysis Note

The biological activity is measured by its ability to bind recombinant mouse ephrin-B2/Fc in an ELISA assay.

Biochem/physiol Actions

Member of Eph receptor tyrosine kinase family shown to bind ephrin-B1 and ephrin-B2; involved in pattern formation and morphogenesis and arterial/venous demarcation of the developing vasculature. EphB4 is also involved in hematopoiesis.

EPHB4 is a member of the Eph family of receptor tyrosine kinases, which play a role in regulating cell adhesion and cell movement during embryonic development. Furthermore, because tissue disorganization and abnormal cell adhesion, movement, and survival characterize the more advanced stages of cancer, the inappropriate functioning of Eph receptor could play a role in malignancy. Furthermore, EPHB4 has been shown to be the major essential interaction partner of Ephrin B2 in angiogenesis.

Other Notes

Extracellular domain of mouse EphB4 (amino acids 1-539) fused to the C-terminal 6X histidine-tagged Fc region of human IgG1 via a polypeptide linker.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

General description

EPHB4 (Ephb1 Eph receptor B4) belongs to EPHB receptor family, which is the biggest family of transmembrane tyrosine kinase receptors. Mammals contain five EPHB receptors, namely, EPHB1-EPHB4 and EPHB6. Their ligands are transmembrane in nature and are called ephrinB1, -B2 and -B3.

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?