EMAIL THIS PAGE TO A FRIEND
E9878 Sigma

Ethyl alcohol-d

Purchase

Properties

isotopic purity   ≥99.5 atom % D

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1170 3/PG 2
Flash Point(F) 
57 °F
Flash Point(C) 
14 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?