EMAIL THIS PAGE TO A FRIEND
E9887 Sigma

Erucic anhydride

~99% (capillary GC)

Synonym: cis-13-Docosenoic anhydride

Purchase

Properties

Related Categories Anhydrides, Biochemicals and Reagents, Fatty Acids and conjugates, Fatty Acyls, Lipids,
InChI Key   LVWSCSGKAPKSBZ-CLFAGFIQSA-N
assay   ~99% (capillary GC)
storage temp.   −20°C

Description

Packaging

Sealed ampule.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?