EMAIL THIS PAGE TO A FRIEND
EMU202651 Sigma

MISSION® esiRNA

esiRNA targeting mouse ENSMUSG00000054351

Purchase

Properties

Related Categories EH-ENSMUSG00000056999, Functional Genomics and RNAi, MISSION esiRNA Technology, Molecular Biology, Mouse esiRNA More...
description   Powered by Eupheria Biotech
concentration   200 ng/μL RNA
esiRNA cDNA target sequence   TCTCACAAACAAGCACATACCACTCTCAGCAGTTGCCCGAAACTTTCGCTTTTTTCCCGTGGTTTTTCACTTTTCAAAGAAACTCAGAGTCACAAGGAAAGCGAAACACCTGTGCGGATGGGCGGCACATGGAGACAGGAAAACACTGCGGCAGGGGAATAAGCGGATTCTTTGGGCGAGAAAGCTGGTGGAGAGGAAAGGCACCCGTGCGGCGCCAGTGCCTGTGGGTGCATATTGGTCCGCACCGTGGGAGAGCCCCTATGCGGGGGCGTGCACATCACTGACTGCACAGGGATCACTCTGCCAGAGGAATGGTGTGTTTTCCTCTTTCCAGAGTTTAACCCAGGCTTTCACGCGGTTCCAGCTTCAACAAGCAGAGCGCCCTGTGACACTCAGCAGACAAACACTTTGGTATTCTGAGACAGCTCAGTTTGTAGAGGACTCACTAGAGAGAATATACAGGAAAGATACCAAAATAACATTCAGGGGA
shipped in   dry ice
storage temp.   −20°C

Description

General description

MISSION esiRNA are endo-ribonuclease prepared siRNA. MISSION esiRNA are a heterogeneous mixture of siRNAs that all target the same mRNA sequence. These multiple silencing triggers lead to highly specific and effective gene silencing.

esiRNA Custom options:
ESISEC: Secondary independent esiRNA for any human or mouse esiRNA
ESIOPEN: Custom esiRNA targeting species specific sequence provided
Contact your local sales rep. or email us at Custom esiRNA for more information.

To view this esiRNA screening tutorial, visit MISSION esiRNA for RNAi Screening in Mammalian Cells.

Physical form

Solution in low-TE (Tris-HCl (10 mM final), EDTA (1 mM final), pH to 8.0.

Legal Information

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?