• USA Home
  • EP1437 - Sigma-Aldrich® VS20 comb
EMAIL THIS PAGE TO A FRIEND
EP1437 Sigma
Sigma-Aldrich® VS20 comb
36 sample MC, thickness 1 mm
Purchase
Description
General description
Comb Prep number indicates the number of preparatory wells. Marker number indicates the number of small volume wells intended for
molecular weight markers. MC indicates multichannel pipette compatible.
Legal Information
Sigma-Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?