• USA Home
  • EXTRA3A - Rabbit ExtrAvidin® Alkaline Phosphatase Staining Kit antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
EXTRA3A Sigma

Rabbit ExtrAvidin® Alkaline Phosphatase Staining Kit antibody produced in goat

Synonym: ExtrAvidin® Alkaline Phosphatase Staining Kits

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibody Kits, ExtrAvidin Staining Kits, Immunohistology Kits,
usage   sufficient for 2,000 tests ELISA, Dot blot
  sufficient for 200 tests Immunohistology
shipped in   wet ice
storage temp.   2-8°C

Description

Components

3 mL ExtrAvidin®-Alkaline Phosphatase
3 mL Biotinylated Secondary Antibodies
Complete Instructions and Assay Protocols

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Use in immunohistology, ELISA, and immunoblotting assays.
Affinity Isolated Antibodies have been adsorbed with human IgG and IgM to minimize cross-reactivity.
Biotinylated antibodies contain a spacer which improves accessibility for the ExtrAvidin® conjugates.

General description

These kits comprise universal reagents for use with primary antibodies in immunohistology, ELISA, and immunoblotting.
ExtrAvidin® is a unique form of avidin, available only from Sigma, that combines the high specificity and affinity of avidin for biotin with low non-specific binding at physiological pH. ExtrAvidin® alkaline phosphatase exhibits high sensitivity with low background.

Legal Information

ExtrAvidin is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?