EMAIL THIS PAGE TO A FRIEND
F0148 Sigma

Fmoc-Ser(tBu)-ODhbt

Synonym: Fmoc-O-tert-butyl-L-serine 3,4-dihydro-4-oxo-1,2,3-benzotriazin-3-ylester

  • CAS Number 109434-27-7

  • Empirical Formula (Hill Notation) C29H28N4O6

  • Molecular Weight 528.56

  •  Beilstein Registry Number 4283373

  •  MDL number MFCD00133600

  •  PubChem Substance ID 24894352

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   NCZXTUHFIKZNNH-VWLOTQADSA-N
impurities   ~10% solvents
storage temp.   −20°C

Description

Packaging

1 g in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?