• USA Home
  • F0254 - D-Fructose 1,6-bisphosphatase from torula yeast

EMAIL THIS PAGE TO A FRIEND
F0254 Sigma

D-Fructose 1,6-bisphosphatase from torula yeast

ammonium sulfate suspension, 10-30 units/mg protein (biuret)

Synonym: D-Fructose-1,6-bisphosphate 1-phosphohydrolase, Hexose diphosphatase

Purchase

Description

Unit Definition

One unit will convert 1.0 μmole of fructose 1,6-diphosphate to fructose 6-phosphate and inorganic phosphate per min at pH 9.5 at 25°C.

Physical form

Suspension in 3.2 M (NH4)2SO4, 5 mM Mg(OAc)2, 0.1 mM EDTA, pH approx. 6.0

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?