• USA Home
  • F0550 - Anti-Flt-3 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
F0550 Sigma

Anti-Flt-3 antibody produced in goat

affinity isolated antibody, lyophilized powder

Purchase

Description

Immunogen

recombinant Flt-3 extracellular domain, expressed in NS0 cells.

Physical form

Lyophilized from 0.2 μm filtered solution of phosphate buffered saline.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?