EMAIL THIS PAGE TO A FRIEND
F0760 Sigma

Potassium formate

Synonym: Formic acid potassium salt

  • CAS Number 590-29-4

  • Linear Formula HCOOK

  • Molecular Weight 84.12

  •  Beilstein Registry Number 3623272

  •  EC Number 209-677-9

  •  MDL number MFCD00013100

Purchase

Properties

InChI Key   WFIZEGIEIOHZCP-UHFFFAOYSA-M
mp   165-168 °C(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

294454 Potassium formate, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?