EMAIL THIS PAGE TO A FRIEND
F0766 Sigma

Fungi TISSUE-TROL

from mouse lung containing Candida albicans

Purchase

Properties

Related Categories Cell Biology, Hematology and Histology, TISSUE-TROL Control Slides More...
Quality Level   GMP-IVD
biological source   from mouse lung containing Candida albicans
shelf life   Expiry date on the label.
IVD   for in vitro diagnostic use

Description

Components

25 slides per box. 1 stained slide is provided with each box. The stained slide has been stained with Sigma Procedure No. HT100.

Application

Suitable for use with Sigma Procedure HT100 (Silver Stain), Modified GMS.

Legal Information

TISSUE-TROL is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

comparable product

Product #

Description

Add to Cart

TTR004 Fungi TISSUE-TROL Control Slides, from mouse lung containing Candida albicans

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?