• USA Home
  • F0777 - Formimino-L-glutamic acid Transferase from porcine liver

EMAIL THIS PAGE TO A FRIEND
F0777 Sigma

Formimino-L-glutamic acid Transferase from porcine liver

lyophilized powder, ≥1 units/mg protein

Synonym: 5-Formiminotetrahydrofolate: L-glutamate N-formiminotransferase, FIGLU Enzymes

Purchase

Description

Unit Definition

One unit will convert 1.0 μmole of FIGLU and THF to L-glutamic acid and 5-formimino-THF per min at pH 7.2 at 25 °C (measured as 5,10-methenyl-THF after perchloric acid treatment).

Physical form

Lyophilized powder containing potassium phosphate buffer salts

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?