• USA Home
  • F1005 - Fast Bordeaux GP salt

EMAIL THIS PAGE TO A FRIEND
F1005 Sigma

Fast Bordeaux GP salt

Synonym: 4-Methoxy-2-nitrobenzenediazonium chloride hemi(zinc chloride) salt

  • Empirical Formula (Hill Notation) C7H6ClN3O3 · 0.5ZnCl2

  • Molecular Weight 283.74

  •  Colour Index Number 37135

  •  MDL number MFCD00135644

  •  PubChem Substance ID 329799671

Purchase

Properties

InChI Key   QFMJFXFXQAFGBO-UHFFFAOYSA-N
composition   Dye content, >90%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?