• USA Home
  • F138 - Anti-FosB antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
F138 Sigma

Anti-FosB antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Gene Regulation, Antibodies to Common Transcription Factors, Antibodies to Transcription Factors,
species reactivity   mouse
application(s)   western blot (chemiluminescent): 10 μg/mL using FosB and FosB2 expressed in mouse fibroblasts NIH3T3 cells following serum stimulation
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
mol wt   antigen mol wt 50-60 kDa
storage temp.   −20°C
Gene Information   mouse ... Fosb(14282)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide (Thr-Ser-Tyr-Ser-Thr-Pro-Gly-Leu-Ser-Ala-Tyr-Ser-Thr-Gly-Gly-Ala), derived from amino acids 102-117 of mouse FosB.

Physical form

Lyophilized from phosphate buffered saline.

Specificity

The antibody reacts specifically with FosB and FosB2 expressed in mouse fibroblasts (NIH 3T3 cells) following serum stimulation. It does not react with FosB in HeLa cells.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Reconstitution

Reconstitute with 100 μl deionized water.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Documents

Certificate of Analysis

Certificate of Origin

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, type N95 (US), type P1 (EN143) respirator filter
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?