EMAIL THIS PAGE TO A FRIEND
F1505 Sigma

Fast Red KB Salt

Synonym: 5-Chloro-2-methylbenzenediazonium 1,5-naphthalenedisulfonate

Purchase

Properties

InChI Key   SGEIBXBCRKDSPV-UHFFFAOYSA-M

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?