• USA Home
  • F155 - Fenoterol hydrobromide

EMAIL THIS PAGE TO A FRIEND
F155 Sigma

Fenoterol hydrobromide

solid

Synonym: 5-[1-Hydroxy-2-[[2-(4-hydroxyphenyl)-1-methylethyl]amino]ethyl]-1,3-benzenediol, Berotec

Purchase

Properties

form   neat
  solid
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
color   white
solubility   H2O: 50 mg/mL
format   neat

Description

Biochem/physiol Actions

β2 Adrenoceptor agonist

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DO1500000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

F1016 Fenoterol hydrobromide

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?