EMAIL THIS PAGE TO A FRIEND
F1630 Sigma

Fast Red RL salt

Synonym: 2-Methyl-4-nitrobenzenediazonium tetrafluoroborate

  • CAS Number 455-90-3

  • Empirical Formula (Hill Notation) C7H6N3O2 · BF4

  • Molecular Weight 250.95

  •  Colour Index Number 37100

  •  EC Number 207-252-2

  •  MDL number MFCD00078520

Purchase

Properties

InChI Key   IEYBUBIALAIMLS-UHFFFAOYSA-N
composition   Dye content, >90%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XT0565000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?