• USA Home
  • F1634 - Ficin Agarose from fig tree latex

EMAIL THIS PAGE TO A FRIEND
F1634 Sigma

Ficin Agarose from fig tree latex

lyophilized powder, 500-1,000 units/g agarose

Purchase

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of BAEE per min at pH 7.0 at 25 °C.

Physical form

Stabilized with lactose.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?