• USA Home
  • F1759 - α-L-(−)-Fucose 1-phosphate bis(cyclohexylammonium) salt

EMAIL THIS PAGE TO A FRIEND
F1759 Sigma

α-L-(−)-Fucose 1-phosphate bis(cyclohexylammonium) salt

~99%

  • CAS Number 24333-03-7

  • Empirical Formula (Hill Notation) C6H11O8P · 2C6H14N

  • Molecular Weight 442.48

  •  MDL number MFCD00057508

  •  PubChem Substance ID 329799686

Purchase

Properties

Related Categories Biochemicals and Reagents, Carbohydrates, Carbohydrates A to Z, Carbohydrates D-F, Monosaccharide More...
InChI Key   FQMPFZHILABVMA-ZXNJZJEFSA-N
assay   ~99%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?