• USA Home
  • F1902 - Monoclonal Anti-HLA-DR−FITC antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
F1902 Sigma

Monoclonal Anti-HLA-DR−FITC antibody produced in mouse

clone HK14, purified immunoglobulin, buffered aqueous solution

Synonym: Monoclonal Anti-HLA-DR

Properties

Related Categories Alphabetical Index, Analytical Cytology, Antibodies, Antibodies for Cell Biology, Antibodies for Immune Cell Signaling and Blood,
species reactivity   human
application(s)   flow cytometry: 10 μL using 1 × 106 cells
clone   HK14, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG2a
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... HLA-DRA(3122), HLA-DRB1(3123)
biological source   mouse
conjugate   FITC conjugate

Description

Immunogen

GM-1500 human tumor cell line

Antigen Background

HLA-DR is located on chromosome 6. It is composed of a heavy and light chain, which are two non-covalently associated subunits of molecular weight 33-36 kDa and 24-30 kDa, respectively. These class II antigens are expressed primarily on B cells, and on monocytes and activated T cells, and function in cell-cell recognition in regulation of the immune response.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 15 mM sodium azide

Specificity

Recognizes HLA-DR, a product of the human major histocompatibility complex. The antibody is specific for a non-polymorphic HLA-DR determinant.

Preparation Note

Prepared by conjugation to fluorescein isothiocyanate isomer I (FITC). This green dye is efficiently excited at 495 nm and emits at 525 nm.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?