• USA Home
  • F2126 - 6-Fluoroindole-3-carboxaldehyde

EMAIL THIS PAGE TO A FRIEND
F2126 Sigma

6-Fluoroindole-3-carboxaldehyde

crystalline

  • CAS Number 2795-41-7

  • Empirical Formula (Hill Notation) C9H6FNO

  • Molecular Weight 163.15

  •  MDL number MFCD00069703

  •  PubChem Substance ID 24894815

Purchase

Properties

Related Categories Aldehydes, Biochemicals and Reagents, Building Blocks, C9, Carbonyl Compounds,
InChI Key   CWCYUOSLRVAKQZ-UHFFFAOYSA-N
form   crystalline
color   yellow
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?