• USA Home
  • F2260 - N-Formyl-L-methionine β-naphthylamide

EMAIL THIS PAGE TO A FRIEND
F2260 Sigma

N-Formyl-L-methionine β-naphthylamide

  • CAS Number 76078-88-1

  • Empirical Formula (Hill Notation) C16H18N2O2S

  • Molecular Weight 302.39

  •  MDL number MFCD00063339

  •  PubChem Substance ID 24894819

Purchase

Properties

Related Categories A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids
InChI Key   SJCDRWNGPZQCPN-HNNXBMFYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?