EMAIL THIS PAGE TO A FRIEND
F2422 Sigma

2× Fixation Buffer

Purchase

Properties

storage temp.   −20°C

Description

Other Notes

20 mm MES, pH 5.6, 600 mM mannitol, 0.6% formaldehyde

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?