• USA Home
  • F2520 - Fentanyl-d5 citrate salt solution

EMAIL THIS PAGE TO A FRIEND
F2520 Sigma

Fentanyl-d5 citrate salt solution

100 μg/mL in methanol

Synonym: N-(Phenyl-d5)-N-(1-[2-phenylethyl]-4-piperidinyl)propanamide citrate salt solution

  • CAS Number 437-38-7

  • Empirical Formula (Hill Notation) C22D5H23N2O · C6H8O7

  • Molecular Weight 533.62

Purchase

Properties

drug control   USDEA Schedule II; Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada
concentration   100 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Quality

Concentrations are based on the equivalent of anhydrous free acid/base, corrected for salt form and water content.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
3
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?