EMAIL THIS PAGE TO A FRIEND
F2635 Sigma

N-Formyl-Met-Met-Met

≥97% (TLC)

  • CAS Number 59881-03-7

  • Empirical Formula (Hill Notation) C16H29N3O5S3

  • Molecular Weight 439.61

  •  MDL number MFCD00037359

  •  PubChem Substance ID 24894828

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chemotactic Peptides, Immunomodulators, Peptides and Proteins,
InChI Key   JAGURBYHFCUGLL-UHFFFAOYSA-N
assay   ≥97% (TLC)
storage temp.   −20°C

Description

Amino Acid Sequence

Nfor-Met-Met-Met

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?